Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bmf Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299460MFV610
Description
Bmf Polyclonal antibody specifically detects Bmf in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
Bmf | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence | |
Bcl2 modifying factor, bcl-2-modifying factor, FLJ00065 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Bmf. | |
0.1 mL | |
Apoptosis, Cancer, Tumor Suppressors | |
90427 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction