Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bmf Antibody (OTI6F3), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272311PCP
Description
Bmf Monoclonal antibody specifically detects Bmf in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Bmf | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
OTI6F3 | |
Western Blot, Immunohistochemistry | |
Bcl2 modifying factor, bcl-2-modifying factor, FLJ00065 | |
Full length human recombinant protein of human BMF ( NP_001003940) produced in E.coli. | |
0.1 mL | |
Apoptosis, Cancer, Tumor Suppressors | |
90427 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction