Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bmf Antibody (SR1375), Novus Biologicals™
SDP

Catalog No. p-200006156 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259260 100 μg
NB259263 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB259260 Supplier Novus Biologicals Supplier No. NBP322130100UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Bmf Monoclonal antibody specifically detects Bmf in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry

Specifications

Antigen Bmf
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone SR1375
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias Bcl2 modifying factor, bcl-2-modifying factor, FLJ00065
Host Species Rabbit
Immunogen A synthesized peptide derived from human Bmf (Uniprot #: Q96LC9)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 90427
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.