Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMF Monoclonal Antibody (OTI7B3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA807513

Description
The protein encoded by this gene belongs to the BCL2 protein family. BCL2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. This protein contains a single BCL2 homology domain 3 (BH3), and has been shown to bind BCL2 proteins and function as an apoptotic activator. This protein is found to be sequestered to myosin V motors by its association with dynein light chain 2, which may be important for sensing intracellular damage and triggering apoptosis. Alternatively spliced transcript variants encoding different isoforms have been identified.Specifications
BMF | |
Monoclonal | |
Unconjugated | |
BMF | |
Bcl2 modifying factor | |
Mouse | |
Affinity Chromatography | |
RUO | |
90427 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
OTI7B3 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q96LC9 | |
BMF | |
Full length human recombit protein of human BMF ( NP_001003940) produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction