Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bmf Monoclonal Antibody (OTI9B7)
GREENER_CHOICE

Catalog No. PIMA526878
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526878 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526878 Supplier Invitrogen™ Supplier No. MA526878
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bmf Monoclonal Antibody for Western Blot, IHC (P)

The protein encoded by this gene belongs to the BCL2 protein family. BCL2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. This protein contains a single BCL2 homology domain 3 (BH3), and has been shown to bind BCL2 proteins and function as an apoptotic activator. This protein is found to be sequestered to myosin V motors by its association with dynein light chain 2, which may be important for sensing intracellular damage and triggering apoptosis. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bmf
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI9B7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BMF
Gene Accession No. Q96LC9
Gene Alias AW260063; AW411872; Bcl2 modifying factor; Bcl-2 modifying factor; bcl-2-modifying factor; Bmf; FLJ00065
Gene Symbols BMF
Host Species Mouse
Immunogen Full length human recombinant protein of BMF produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90427
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.