Learn More
BMF Mouse anti-Human, Clone: OTI3E5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF807460

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene belongs to the BCL2 protein family. BCL2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. This protein contains a single BCL2 homology domain 3 (BH3), and has been shown to bind BCL2 proteins and function as an apoptotic activator. This protein is found to be sequestered to myosin V motors by its association with dynein light chain 2, which may be important for sensing intracellular damage and triggering apoptosis. Alternatively spliced transcript variants encoding different isoforms have been identified.Specifications
BMF | |
Monoclonal | |
Unconjugated | |
BMF | |
Bcl2 modifying factor | |
Mouse | |
Affinity Chromatography | |
RUO | |
90427 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3E5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96LC9 | |
BMF | |
Full length human recombit protein of human BMF ( NP_001003940) produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.