Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Bmf Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110759
Description
Immunogen sequence: VEELEDDVFQ PEDGEPVTQP GSLLSADLFA QSLLDCPLSR LQLFPLTHCC GPGLRPTSQE DKATQTLSPA SPSQGVMLPC GVTEEPQRLF YGNAGYRLPL PASFPAVLPI GEQPPEGQWQ HQAEVQIARK LQCIADQFHR LHVQQHQQNQ.
The protein encoded by this gene belongs to the BCL2 protein family. BCL2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. This protein contains a single BCL2 homology domain 3 (BH3), and has been shown to bind BCL2 proteins and function as an apoptotic activator. This protein is found to be sequestered to myosin V motors by its association with dynein light chain 2, which may be important for sensing intracellular damage and triggering apoptosis. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| Bmf | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96LC9 | |
| BMF | |
| Recombinant Protein corresonding to Human Bmf. Recombinant protein control fragment (Product #RP-88647). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| BMF | |
| AW260063; AW411872; Bcl2 modifying factor; Bcl-2 modifying factor; bcl-2-modifying factor; Bmf; FLJ00065 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 90427 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction