Learn More
Description
Specifications
Specifications
| Antigen | beta Amyloid |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | MOAB-2 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:5000, ELISA 1:100-1:1000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:500, Immunoprecipitation 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, Immunohistochemistry-Frozen, Immunoblotting reported in scientific literature (PMID 28314768), Dot Blot reported in scientific literature (PMID 22423893), Immunohistochemistry Free-Floating reported in scientific literature (PMID 25747037) |
| Gene Accession No. | P05067 |
| Gene Alias | Abeta, Amyloid beta, beta-amyloid peptide |
| Gene Symbols | APP |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
