Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BMI-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143177
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143177 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5143177 Supplier Invitrogen™ Supplier No. PA5143177
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

Component of the Polycomb group (PcG) multiprotein PRC1 complex, a complex required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development. PcG PRC1 complex acts via chromatin remodeling and modification of histones; it mediates monoubiquitination of histone H2A 'Lys-119', rendering chromatin heritably changed in its expressibility. In the PRC1 complex, it is required to stimulate the E3 ubiquitin-protein ligase activity of RNF2/RING2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMI-1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene BMI1
Gene Accession No. P35226
Gene Alias AW546694; B lymphoma Mo-MLV insertion region 1; B lymphoma Mo-MLV insertion region 1 homolog; Bmi1; Bmi-1; BMI1 polycomb ring finger oncogene; BMI1 polycomb ring finger proto-oncogene; BMI1 proto-oncogene, polycomb ring finger; FLVI2/BMI1; flvi-2/bmi-1; murine leukemia viral (bmi-1) oncogene homolog; Pcgf4; Pcgf4 antibody; Polycomb complex protein BMI-1; polycomb group protein Bmi1; polycomb group ring finger 4; polycomb group RING finger protein 4; RING finger protein 51; RNF51; RP11-573G6.1
Gene Symbols BMI1
Host Species Goat
Immunogen Peptide with sequence C-NHQSSFANRPRKS.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 648
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.