Learn More
Invitrogen™ BMI-1 Polyclonal Antibody, HRP

Description
Suggested positive control: human and mouse skeletal muscle. Immunogen sequence homology: primate, feline, chicken, bovine, rabbit, rat and Zebrafish protein (all 92%).
Specifications
Specifications
| Antigen | BMI-1 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.09 mg/mL |
| Conjugate | HRP |
| Formulation | PBS with no preservative |
| Gene | BMI1 |
| Gene Accession No. | P25916, P35226 |
| Gene Alias | AW546694; B lymphoma Mo-MLV insertion region 1; B lymphoma Mo-MLV insertion region 1 homolog; Bmi1; Bmi-1; BMI1 polycomb ring finger oncogene; BMI1 polycomb ring finger proto-oncogene; BMI1 proto-oncogene, polycomb ring finger; FLVI2/BMI1; flvi-2/bmi-1; murine leukemia viral (bmi-1) oncogene homolog; Pcgf4; Pcgf4 antibody; Polycomb complex protein BMI-1; polycomb group protein Bmi1; polycomb group ring finger 4; polycomb group RING finger protein 4; RING finger protein 51; RNF51; RP11-573G6.1 |
| Gene Symbols | BMI1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.