Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMP-1/PCP Antibody (OTI3E9), Alexa Fluor™ 647, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270255AF647

Catalog No. N270255A647


Only null left
Add to Cart

Description

Description

BMP-1/PCP Monoclonal specifically detects BMP-1/PCP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

BMP-1/PCP
Monoclonal
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
BMP1
Recombinant protein fragment corresponding to amino acids 225-617 of human BMP-1/PCP (NP_001190) produced in E.coli.
0.1 mL
Primary
Human, Mouse, Rat
IgG1
Western Blot, Immunocytochemistry, Immunofluorescence
OTI3E9
Western Blot, Immunocytochemistry/Immunofluorescence
BMP-1, bone morphogenetic protein 1, EC 3.4.24, EC 3.4.24.19, FLJ44432, Mammalian tolloid protein, mTld, PCOLC, PCP, PCP2, procollagen C-endopeptidase, Procollagen C-proteinase, TLD
Mouse
Affinity Purified
Cytokine Research
649.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.