Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMP-1/PCP Antibody (OTI3E9), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP246579
Description
Ensure accurate, reproducible results in Western Blot, Immunofluorescence
BMP-1/PCP Monoclonal specifically detects BMP-1/PCP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunofluorescence.Specifications
| BMP-1/PCP | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:4000, Immunocytochemistry/Immunofluorescence 1:100 | |
| BMP-1, bone morphogenetic protein 1, EC 3.4.24, EC 3.4.24.19, FLJ44432, Mammalian tolloid protein, mTld, PCOLC, PCP, PCP2, procollagen C-endopeptidase, Procollagen C-proteinase, TLD | |
| Mouse | |
| 80.5 kDa | |
| 0.1 mL | |
| Cytokine Research | |
| 649 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI3E9 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| BMP1 | |
| Human recombinant protein fragment corresponding to amino acids 225-617 of human BMP1(NP_001190) produced in E.coli. | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction