Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMP-10 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$464.00
Specifications
Antigen | BMP-10 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.5 - 4 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
BMP-10 Polyclonal specifically detects BMP-10 in Human samples. It is validated for Western Blot.Specifications
BMP-10 | |
Western Blot 0.5 - 4 ug/mL | |
Polyclonal | |
Rabbit | |
Cell Cycle and Replication | |
O95393 | |
27302 | |
E. coli expressed human BMP-10 | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
BMP-10, bone morphogenetic protein 10, MGC126783 | |
BMP10 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title