Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMP-15/GDF-9B Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281743
Description
BMP-15/GDF-9B Polyclonal specifically detects BMP-15/GDF-9B in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
BMP-15/GDF-9B | |
Polyclonal | |
Unconjugated | |
PBS | |
BMP15 | |
Rabbit polyclonal BMP15 antibody was raised against an 18 amino acid peptide near the amino terminus of human BMP15. The immunogen is located within amino acids 60 - 110 of BMP15. Amino Acid Squence: LYRRSADSHGHPRENRTI | |
Affinity Purified | |
RUO | |
9210 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
BMP-15, bone morphogenetic protein 15, GDF-9B, Growth/differentiation factor 9B, ODG2, POF4 | |
Rabbit | |
43 kDa | |
0.1 mg | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction