Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BMP-15 Recombinant Rabbit Monoclonal Antibody (6K5C3)
GREENER_CHOICE

Catalog No. PIMA535450
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535450 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535450 Supplier Invitrogen™ Supplier No. MA535450
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: LHPFQISFRQ LGWDHWIIAP PFYTPNYCKG TCLRVLRDGL NSPNHAIIQN LINQLVDQSV PRPSCVPYKY VPISVLMIEA NGSILYKEYE GMIAESCTCR.

BMP15, or GDF9B is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily that are involved in embryonic development and adult tissue homeostasis. BMP15 is expressed exclusively in the oocyte. It is an oocyte-specific growth/differentiation factor that may be involved in oocyte maturation and follicular development. Defects in BMP15 are the cause of ovarian dysgenesis type 2. BMP15 could be used as an oogenesis marker to track human amniotic fluid stem cells differentiation into the oocyte-like cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMP-15
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 6K5C3
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BMP15
Gene Accession No. O95972, Q9Z0L4
Gene Alias AU015375; AU018861; AU021453; BMP; BMP15; Bmp-15; Bone morphogenetic protein; Bone morphogenetic protein 15; C86824; C87336; GDF9B; GDF-9B; growth differentiation factor-9B; growth/differentiation factor 9B; ODG2; POF4
Gene Symbols BMP15
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 293-392 of human BMP15 (O95972).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12155, 59302, 9210
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.