Learn More
Invitrogen™ BMP-15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595615
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human placenta tissue, human Caco-2 whole cell, rat lung tissue, mouse lung tissue. IHC: mouse lung tissue, human gastric cancer tissue, human intestinal cancer tissue, human lung cancer tissue, human ovary cancer tissue, human placenta tissue, human prostatic cancer tissue, human thyroid cancer tissue, human tonsil tissue, rat lung tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
BMP15, or GDF9B is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily that are involved in embryonic development and adult tissue homeostasis. BMP15 is expressed exclusively in the oocyte. It is an oocyte-specific growth/differentiation factor that may be involved in oocyte maturation and follicular development. Defects in BMP15 are the cause of ovarian dysgenesis type 2. BMP15 could be used as an oogenesis marker to track human amniotic fluid stem cells differentiation into the oocyte-like cells.
Specifications
| BMP-15 | |
| Polyclonal | |
| Unconjugated | |
| BMP15 | |
| AU015375; AU018861; AU021453; BMP; BMP15; Bmp-15; Bone morphogenetic protein; Bone morphogenetic protein 15; C86824; C87336; GDF9B; GDF-9B; growth differentiation factor-9B; growth/differentiation factor 9B; ODG2; POF4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12155, 59302, 9210 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| O95972, Q9Z0L4 | |
| BMP15 | |
| A synthetic peptide corresponding to a sequence of human BMP15 (KGTCLRVLRDGLNSPNHAIIQNLINQLVDQ). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.