Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMP-2 Antibody (15B3), Alexa Fluor™ 594, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP242651AF594

Catalog No. NP24261A94


Only null left
Add to Cart

Description

Description

BMP-2 Monoclonal specifically detects BMP-2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

BMP-2
Monoclonal
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
BMP2
Recombinant human BMP2 (283-396aa) purified from E. coli
Primary
Human
IgG2b κ
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
15B3
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2
Mouse
100 μL
650
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.