Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMP-2 Antibody (9E10G12) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$560.50
Specifications
Antigen | BMP-2 |
---|---|
Clone | 9E10G12 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
Description
BMP-2 Monoclonal specifically detects BMP-2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
BMP-2 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Biologically Active Proteins, Cytokine Research, Embryonic Stem Cell Markers, Immunology, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers, Stem Cell Signaling Pathway | |
PBS with 0.05% Sodium Azide | |
650 | |
Purified recombinant fragment of human BMP2 (AA: 283-396) expressed in E. Coli. | |
Primary | |
44.7 kDa |
9E10G12 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2 | |
BMP2 | |
IgG1 | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title