Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ BMP-2 Quantikine ELISA Kit
GSA_VA

Catalog No. p-4753748
Click to view available options
Quantity:
1 Plate
6 Plates
This item is not returnable. View return policy
This item is not returnable. View return policy

BMP-2 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The BMP-2 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human, mouse, rat BMP-2 in cell culture supernates (50 ul), bone extracts (50 ul).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Assay Range 62.50 - 4000pg/mL
Assay Sensitivity 29pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. Cross-reactivity observed with 1 or more available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Bone Extracts, Cell Culture Supernatant
Specificity Natural and mammalian-expressed recombinant human BMP-2. This assay does not recognize E. coli-expressed BMP-2.
Sufficient For 1 x 96-well Plate
Target Species Human, Mouse, Rat
For Use With (Application) ELISA
Gene ID (Entrez) 650 (Human); 12156 (Mouse); 29373 (Rat)
Quantity 1 Plate
Sample Volume Cell Culture Supernates (50uL),Bone Extracts (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym BDA2, BMP2, BMP-2, BMP-2 A, BMP2ABone morphogenetic protein 2 A, bone morphogenetic protein 2, SSFSC
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.