Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BMP-3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550921
Description
BMP3 Polyclonal Antibody detects endogenous levels of BMP3 protein.
The BMPs belong to the TGF-Beta superfamilyBMPs stimulate the production of bone matrix proteins and thereby alter stromal cell and osteoclast proliferation, a key step in bone marrow development. In addition to promoting ectopic bone and cartilage development, BMPs regulate the growth, differentiation, chemotaxis, proliferation, and apoptosis of various cell types, including mesenchymal cells, epithelial cells, hematopoietic cells, and neuronal cells. BMPs also play a role in dorsal/ventral patterning. BMP3, highly expressed in lung, ovary and small intestine, participates in cartilage and bone formation. BMP3 and BMP2 genes map to conserved regions between human and mouse.
Specifications
BMP-3 | |
Polyclonal | |
Unconjugated | |
BMP3 | |
9130206H07; 9530029I04Rik; BMP; Bmp3; Bmp-3; BMP3A; BMP-3A; Bone morphogenetic protein; Bone morphogenetic protein 3; bone morphogenetic protein 3 (osteogenic); bone morphogenetic protein 3A; bone morphogenetic protein-3; osteogenin; PBMP3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
110075, 25667, 651 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P12645, P49002, Q8BHE5 | |
BMP3 | |
Synthesized peptide derived from the Internal region of human BMP-3. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction