Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMP-3b/GDF-10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP18857425UL
Description
BMP-3b/GDF-10 Polyclonal specifically detects BMP-3b/GDF-10 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
BMP-3b/GDF-10 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
BIP, BMP-3b, BMP3B, bone morphogenetic protein 3B, Bone-inducing protein, GDF-10, growth differentiation factor 10, Growth/differentiation factor 10 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
GDF10 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:ISEPNSVAVTLQRYDPFPAGDPEPRAAPNNSADPRVRRAAQATGPLQDNELPGLDERPPRAHAQHFHKHQLWPSPFRALKPRPGRKDRRKKGQEVFMAASQVLDFDEKTMQKA | |
25 μL | |
Cytokine Research | |
2662 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction