Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMP-4 Antibody (OTI6B7), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270256IR
Description
BMP-4 Monoclonal specifically detects BMP-4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
BMP-4 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
BMP4 | |
Recombinant fragment expressed in E.coli corresponding to amino acids 293-408 of human BMP-4. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI6B7 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
BMP-2B, BMP2B1, BMP2BMCOPS6, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, OFC11, ZYME | |
Mouse | |
Affinity Purified | |
Biologically Active Proteins, Embryonic Stem Cell Markers, Stem Cell Markers, Stem Cell Signaling Pathway | |
652.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction