Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMP-4 Antibody (OTI6B7), Janelia Fluor™ 549, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270256JF549

Catalog No. N270256J549


Only null left
Add to Cart

Description

Description

BMP-4 Monoclonal specifically detects BMP-4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

BMP-4
Monoclonal
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
BMP4
Recombinant fragment expressed in E.coli corresponding to amino acids 293-408 of human BMP-4.
0.1 mL
Primary
Human, Mouse, Rat
IgG1
Western Blot, Immunocytochemistry, Immunofluorescence
OTI6B7
Western Blot, Immunocytochemistry/Immunofluorescence
BMP-2B, BMP2B1, BMP2BMCOPS6, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, OFC11, ZYME
Mouse
Affinity Purified
Biologically Active Proteins, Embryonic Stem Cell Markers, Stem Cell Markers, Stem Cell Signaling Pathway
652.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.