Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech BMP-4 Monoclonal Antibody, PeproTech®

Catalog No. 501626189
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-162-6189 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-162-6189 Supplier PeproTech Supplier No. 500M121500UG
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

AA Sequence of recombinant protein: HHSQRARKKN KNCRRHSLYV DFSDVGWNDW IVAPPGYQAF YCHGDCPFPL ADHLNSTNHA IVQTLVNSVN SSIPKACCVP TELSAISMLY LDEYDKVVLK NYQEMVVEGC GCR. Preparation: Produced in mice immunized with highly pure Recombinant Human BMP-4. Anti-Human BMP-4-specific antibody was purified from cell culture by Protein G affinity chromatography.

The protein encoded by this gene is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. This particular family member plays an important role in the onset of endochondral bone formation in humans, and a reduction in expression has been associated with a variety of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva. Alternative splicing in the 5' untranslated region of this gene has been described and three variants are described, all encoding an identical protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMP-4
Applications ELISA, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation PBS with no preservative
Gene BMP4
Gene Accession No. P12644
Gene Alias BMP; BMP2B; BMP-2B; Bmp2b1; Bmp2b-1; Bmp4; Bmp-4; BOMPR4A; Bone morphogenetic protein; Bone morphogenetic protein 2B; bone morphogenetic protein 4; bone morphogenetic protein 4 preproprotein; DVR4; Dvr-4; MCOPS6; OFC11; ZYME
Gene Symbols BMP4
Host Species Mouse
Immunogen HeLa cells-derived Recombinant Human BMP-4.
Purification Method Protein G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 652
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.