Learn More
Invitrogen™ BMP-5 Polyclonal Antibody
Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA547821
Description
In Western blots, approximately 20% cross-reactivity with recombinant human BMP-5 is observed. In direct ELISAs, approximately 100% cross-reactivity with recombinant human BMP-5 is observed, and approximately 4% cross-reactivity with recombinant mouse (rm) BMP-6 and rmBMP-7 is observed, and less than 1% cross-reactivity with rmBMP-8b is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
TGF-β family members are key modulators of cell proliferation, differentiation, matrix synthesis, and apoptosis. As implied by their name, BMPs initiate, promote, and regulate the development, growth, and remodeling of bone and cartilage. In addition to this role, BMPs are also involved in prenatal development and postnatal growth, remodeling, and maintenance of a variety of other tissues and organs. BMP-5 is expressed in the nervous system, lungs and liver. It is a known regulator for dendritic growth in sympathetic neurons. BMP-5 is a 454 amino acid precursor protein that is cleaved to release the biologically active C-terminal mature protein.
Specifications
BMP-5 | |
Polyclonal | |
Unconjugated | |
Bmp5 | |
AU023399; BMP; BMP 5; Bmp5; BMP-5; bone morphogenenic protein-5; Bone morphogenetic protein; bone morphogenetic protein 5; MGC34244; se; short ear | |
Sheep | |
Antigen Affinity Chromatography | |
RUO | |
12160 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Frozen), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P49003 | |
Bmp5 | |
CHO-derived recombinant mouse BMP-5 Ala315-His452. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.