Learn More
Brady™ BMP™61, BMP™71 Series MetaLabel™ Solvent Resistant Metallized Polyester Label Supply - Light Gray
Metallized Polyester (B-428) labels are designed for applications that use barcodes, alphanumerics, graphic symbols and logos and require nameplate-like quality.
Supplier: Brady™ M61C500428
Description
- Matte, light gray finish gives labels a rating and serial plate appearance
- Labels are designed to withstand numerous solvents and variable temperatures when applied to various surfaces
- RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
- UL approved
- CSA accepted
- Service temperature of -40 to 248° F (-40 to 120° C)
Specifications
Light Gray | |
BMP™61 Labeling System, BMP™71 Label Printer |