Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMP-7 Antibody (4E7), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP104259AF532
Description
BMP-7 Monoclonal specifically detects BMP-7 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
BMP-7 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
BMP7 | |
Recombinant human BMP7 (293-431 aa) purified from E. coli | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
40000000 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
BMP-7, bone morphogenetic protein 7, Eptotermin alfa, OP-1OP1, Osteogenic protein 1 | |
Mouse | |
Protein G purified | |
Biologically Active Proteins, Cytokine Research | |
655.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction