Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMP-7 Antibody (4E7), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP104259FR
Description
BMP-7 Monoclonal specifically detects BMP-7 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
BMP-7 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
BMP7 | |
Recombinant human BMP7 (293-431 aa) purified from E. coli | |
0.1 mL | |
Biologically Active Proteins, Cytokine Research | |
655 | |
Human | |
Purified |
Western Blot, ELISA | |
40000000 | |
Western Blot, ELISA | |
BMP-7, bone morphogenetic protein 7, Eptotermin alfa, OP-1OP1, Osteogenic protein 1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human BMP7 mAb, clone 4E7, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human BMP7 protein. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction