Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMP-7 Antibody (SR1850), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321718100UL
Description
BMP-7 Monoclonal antibody specifically detects BMP-7 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
BMP-7 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunoprecipitation | |
SR1850 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
BMP-7, bone morphogenetic protein 7, Eptotermin alfa, OP-1OP1, Osteogenic protein 1 | |
A synthesized peptide derived from human BMP-7 (Uniprot #: P18075) | |
100 μg | |
Biologically Active Proteins, Cytokine Research | |
655 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction