Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BMP-7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587488
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587488 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587488 Supplier Invitrogen™ Supplier No. PA587488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PHLQGKHNSA PMFMLDLYNA MAVEEGGGPG GQGFSYPYKA VFSTQGPPLA SLQDSHFLTD ADMVMSFVNL VEHDKEFFHP RYHHREFRFD LSKIPEGEAV TAAEFRIYKD YIRERFDNET FRISVYQVLQ EHLGRESDLF LLDSRTLWAS EEGWLVFDIT ATSNHWVVNP RHNLGLQLSV ET Positive Samples: MCF-7, Mouse kidney.

The bone morphogenetic proteins (BMPs) are a family of secreted signaling molecules that can induce ectopic bone growth. Many BMPs are part of the transforming growth factor-beta (TGFB) superfamily. BMPs were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extra skeletal site. Based on its expression early in embryogenesis, BMP7 has a proposed role in early development. In addition, the fact that BMP7 is closely related to BMP5 and BMP6 has lead to speculation of possible bone inductive activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMP-7
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene Bmp7
Gene Accession No. P18075, P23359
Gene Alias BMP; BMP7; BMP-7; bone moorphogenic protein-7; Bone morphogenetic protein; bone morphogenetic protein 7; bone morphogenetic protein 7 (osteogenic protein 1); bone morphogenetic protein 7 preproprotein; bone morphogenic protein 7; Eptotermin alfa; OP1; OP-1; Osteogenic protein 1; RP11-560A15.5
Gene Symbols Bmp7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 70-251 of BMP7 (NP_0017101).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12162, 655, 85272
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.