Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BMP-7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA511720
Description
This antibody is predicted to react with mouse based on sequence homology.
The bone morphogenetic proteins (BMPs) are a family of secreted signaling molecules that can induce ectopic bone growth. Many BMPs are part of the transforming growth factor-beta (TGFB) superfamily. BMPs were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extra skeletal site. Based on its expression early in embryogenesis, BMP7 has a proposed role in early development. In addition, the fact that BMP7 is closely related to BMP5 and BMP6 has lead to speculation of possible bone inductive activity.
Specifications
BMP-7 | |
Polyclonal | |
Unconjugated | |
Bmp7 | |
BMP; BMP7; BMP-7; bone moorphogenic protein-7; Bone morphogenetic protein; bone morphogenetic protein 7; bone morphogenetic protein 7 (osteogenic protein 1); bone morphogenetic protein 7 preproprotein; bone morphogenic protein 7; Eptotermin alfa; OP1; OP-1; Osteogenic protein 1; RP11-560A15.5 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
655 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P18075 | |
Bmp7 | |
KLH conjugated synthetic peptide between 15-44 amino acids from the N-terminal region of human Bmp7. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction