Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BMP-8A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528849
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, Raji. Predicted reactivity: Mouse (100%), Rat (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Bone morphogenic proteins (BMPs) are members of the TGF beta superfamily. BMPs are involved in the induction of cartilage and bone formation.BMP-8 is thought to be involved in early development, as detectable expression has not been found in adult organs. Two BMP-8 proteins exist, namely BMP-8A and BMP-8B (also designated OP-2), and are encoded by two distinct genes.
Specifications
BMP-8A | |
Polyclonal | |
Unconjugated | |
BMP8A | |
BMP; Bmp7r1; Bmp8; Bmp-8; BMP8A; BMP-8A; BMP8B; BMP-8B; Bone morphogenetic protein; Bone morphogenetic protein 8A; OP2; OP-2; Osteogenic protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12163, 353500 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.38 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P34821, Q7Z5Y6 | |
BMP8A | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human BMP8A. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction