Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BMP-8A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA536435
Description
This antibody detects endogenous protein at a molecular weight of 45 kDa. It may cross-react with BMP-8B protein. Purity is >95% by SDS-PAGE.
Bone morphogenic proteins (BMPs) are members of the TGF beta superfamily. BMPs are involved in the induction of cartilage and bone formation.BMP-8 is thought to be involved in early development, as detectable expression has not been found in adult organs. Two BMP-8 proteins exist, namely BMP-8A and BMP-8B (also designated OP-2), and are encoded by two distinct genes.
Specifications
BMP-8A | |
Polyclonal | |
Unconjugated | |
BMP8A | |
BMP; Bmp7r1; Bmp8; Bmp-8; BMP8A; BMP-8A; BMP8B; BMP-8B; Bone morphogenetic protein; Bone morphogenetic protein 8A; OP2; OP-2; Osteogenic protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12163, 353500, 680931 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P34821, Q7Z5Y6 | |
BMP8A | |
Synthetic peptide corresponding to amino acids 264-321 of human BMP-8A. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction