Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BMP-8A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA551103
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA551103 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA551103 Supplier Invitrogen™ Supplier No. PA551103
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BMP8A Polyclonal Antibody detects endogenous levels of BMP8A protein.

Bone morphogenic proteins (BMPs) are members of the TGF beta superfamily. BMPs are involved in the induction of cartilage and bone formation.BMP-8 is thought to be involved in early development, as detectable expression has not been found in adult organs. Two BMP-8 proteins exist, namely BMP-8A and BMP-8B (also designated OP-2), and are encoded by two distinct genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMP-8A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene BMP8A
Gene Accession No. P34821, Q7Z5Y6
Gene Alias BMP; Bmp7r1; Bmp8; Bmp-8; BMP8A; BMP-8A; BMP8B; BMP-8B; Bone morphogenetic protein; Bone morphogenetic protein 8A; OP2; OP-2; Osteogenic protein 2
Gene Symbols BMP8A
Host Species Rabbit
Immunogen Synthesized peptide derived from the Internal region of human BMP-8A (aa 241-290).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12163, 353500, 680931
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.