Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMP-8b Antibody (13E6), DyLight 680, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP242693FR

Catalog No. NBP242693FR


Only null left
Add to Cart

Description

Description

BMP-8b Monoclonal specifically detects BMP-8b in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

BMP-8b
Monoclonal
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
BMP8B
Recombinant human BMP8b (264-402aa) purified from E. coli
100 μL
Cytokine Research
656
Store at 4C in the dark.
IgG2a κ
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
13000000
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
BMP-8, bone morphogenetic protein 8b, MGC131757
Mouse
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.