Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMP-8b Antibody (13E6), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242693IR
Description
BMP-8b Monoclonal specifically detects BMP-8b in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
BMP-8b | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
BMP8B | |
Recombinant human BMP8b (264-402aa) purified from E. coli | |
100 μL | |
Cytokine Research | |
656 | |
Store at 4C in the dark. | |
IgG2a κ |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
13000000 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
BMP-8, bone morphogenetic protein 8b, MGC131757 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction