Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMP-8b Antibody, Novus Biologicals™
SDP

Catalog No. p-7200857 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018173 0.025 mg
NB300698005 0.05 mg
NB300698 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB018173 Supplier Novus Biologicals Supplier No. NB3006980.025MG
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BMP-8b Polyclonal specifically detects BMP-8b in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen BMP-8b
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.50 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, ELISA 1:2000
Formulation Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide
Gene Alias BMP-8, bone morphogenetic protein 8b, MGC131757
Gene Symbols BMP8B
Host Species Rabbit
Immunogen Synthetic peptide: LQTLRAGDEGWLVL conjugated to KLH, corresponding to amino acids 184-197 of BMP8.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 656
Test Specificity BMP8
Target Species Rat, Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.