Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMP-8b Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$482.50

Specifications

Antigen BMP-8b
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB126255
SDP
View Documents
Novus Biologicals
NBP310677100UL
100 μg
Each of 1 for $482.50
Add to Cart
 
Description

Description

BMP-8b Polyclonal specifically detects BMP-8b in Human samples. It is validated for Western Blot.
Specifications

Specifications

BMP-8b
Western Blot
Unconjugated
Rabbit
Cytokine Research
PBS buffer, 2% sucrose
656
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1.0 ug/ml
Polyclonal
Purified
RUO
Human
BMP-8, bone morphogenetic protein 8b, MGC131757
The immunogen is a synthetic peptide directed towards the middle region of human BMP-8b (NP_001711.2). Peptide sequence VRPLRRRQPKKTNELPQANRLPGIFDDVHGSHGRQVCRRHELYVSFQDLG
Affinity purified
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit