Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BMP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582506
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582506 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582506 Supplier Invitrogen™ Supplier No. PA582506
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSEPLNYKDP CKAAAFLGDI ALDEEDLRAF QVQQAVDLRR HTARKSSIKA AVPGNTSTPS CQSTNGQPQR GACGRWRGRS RSRRAATSRP ERVWPDGVIP FVIGGNFTGS Highest antigen sequence indentity to the following orthologs: Rat - 80%, Mouse - 80%.

This gene encodes a protein that is capable of inducing formation of cartilage in vivo. Although other bone morphogenetic proteins are members of the TGF-beta superfamily, this gene encodes a protein that is not closely related to other known growth factors. This gene is expressed as alternatively spliced variants that share an N-terminal protease domain but differ in their C-terminal region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BMP1
Gene Accession No. P13497
Gene Alias BMP; Bmp1; BMP-1; Bone morphogenetic protein; Bone morphogenetic protein 1; FLJ44432; mammalian tolloid protein; mTld; OI13; PCOLC; Pcp; PCP2; pCP-2; procollagen C-endopeptidase; procollagen C-proteinase; procollagen C-proteinase 3; TLD
Gene Symbols BMP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BMP1. Recombinant protein control fragment (Product #RP-90828).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 649
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.