Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMP1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA547222
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547222 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547222 Supplier Thermo Scientific Supplier No. PA547222

Goat Polyclonal Antibody

In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) TLL-2 is observed, and less than 1% cross-reactivity with rhBMP-2, -3, -3b, -4, -5, -6, -7, -8, -9, -10, and -15 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

BMP1 Polyclonal antibody specifically detects BMP1 in Human samples. It is validated for Immunoprecipitation, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMP1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene BMP1
Gene Accession No. P13497
Gene Alias BMP; Bmp1; BMP-1; Bone morphogenetic protein; Bone morphogenetic protein 1; FLJ44432; mammalian tolloid protein; mTld; OI13; PCOLC; Pcp; PCP2; pCP-2; procollagen C-endopeptidase; procollagen C-proteinase; procollagen C-proteinase 3; TLD
Gene Symbols BMP1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human BMP-1/PCP Ala121-Gln730
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 649
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.