Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMP10 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989074
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9074 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9074 Supplier Bioss Supplier No. BS9447R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BMP10 is a member of the TGF-beta family of growth factors. Data suggest that the similar protein in mouse plays an important role in trabeculation of the embryonic heart. In human, this protein may signal through receptor serine/threonine kinases.

Specifications

Antigen BMP10
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene BMP10
Gene Accession No. O95393
Gene Alias BMP; Bmp10; BMP-10; bmp10 {ECO:0000312; bmp10b; Bone morphogenetic protein; Bone morphogenetic protein 10; BOS_11783; EMBL:BAE16990.1, ECO:0000312; RGD:1562986}; si:dkey-13n15.5
Gene Symbols BMP10
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human BMP10.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27302
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.