Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMP2 Mouse anti-Human, Clone:9E10G12, Abnova™
Mouse monoclonal antibody raised against recombinant human BMP2.
Supplier: Abnova Corporation MAB16724
Description
The protein encoded by this gene belongs to the transforming growth factor-beta (TGFB) superfamily. The encoded protein acts as a disulfide-linked homodimer and induces bone and cartilage formation. [provided by RefSeq]Specifications
BMP2 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry Immunocytochemistry (1:200-1:1000) Flow Cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
|
BMP2 | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunocytochemistry, Western Blot | |
9E10G12 | |
bone morphogenetic protein 2 | |
In PBS (0.05% sodium azide) | |
BMP2A | |
Recombinant protein corresponding to amino acid 283-396 of human BMP2 from E. coli. | |
650 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction