Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMP2 Mouse anti-Human, Porcine, Clone: 1C5C2, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 663831IG150UL

Description
Bone Morphogenic Proteins (BMP) are members of the TGF-beta superfamily that affect bone and cartilage formation (Hogan 1996, Reddi 1998 and Francis-West et al. 1999). Mature BMPs are 30-38 kDa proteins that assume a TGF-beta -like cysteine knot configuration. Lovostatin increases bone formation by turning on the bmp-2 gene (Mundy et al. 1999). BMPs stimulate the production of specific bone matrix proteins and alter stromal cell and osteoclast proliferation (Macias et al. 1999, Lecanda et al. 1997). BMPs may also be an important factor for development of the viscera, with roles in cell proliferation, apoptosis, differentiation, and morphogenesis (Hogan 1996, Dale and Wardle 1999). BMPs appear to be responsible for normal dorsal/ventral patterning. Like TGF-beta, BMPs bind to a type II receptor, which then recruits the transducing type I receptor unit, activating the Smad protein signaling pathway (Massague 1994, Derynck 1997, Attisano 1993).Specifications
BMP2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P12643 | |
BMP2 | |
BMP2 Fusion Protein Ag13501 | |
150 μL | |
Primary | |
Human, Pig | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
1C5C2 | |
Unconjugated | |
BMP2 | |
BMP 2, BMP 2A, BMP2, BMP2A, bone morphogenetic protein 2, Bone morphogenetic protein 2A | |
Mouse | |
Protein A | |
RUO | |
100157103, 650 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction