Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMP2 Rabbit anti-Human, Mouse, Non-human Primate, Porcine, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc BMP201AP

Description
BMP2 Polyclonal antibody specifically detects BMP2 in Human, Mouse, Non-human Primate, Porcine samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western BlotSpecifications
BMP2 | |
Polyclonal | |
Unconjugated | |
BMP2 | |
2610024H22Rik; AI467020; AL117858; AW546137; BB189135; BDA2; BMP; BMP2; BMP-2; BMP2A; BMP-2A; BMPRII; BMPR-II; Bone morphogenetic protein; Bone morphogenetic protein 2; bone morphogenetic protein 2 precursor (BMP-2) (BMP-2A); bone morphogenetic protein 2A; H-BMP-2; morphogen | |
Synthetic peptide corresponding to Mouse BMP2 amino acid region 45-60 conjugated to Keyhole Limpet Haemocyanin (KLH). Sequence:SEDVLSEFELRLLSMF | |
200 μL | |
Primary | |
Human, Mouse, Non-human Primate, Pig | |
Antibody | |
IgG |
ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
P12643, P21274 | |
Rabbit | |
Affinity chromatography | |
RUO | |
100157103, 12156, 458090, 650 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction