Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMP2K Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $499.50
Specifications
Antigen | BMP2K |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP1529020
![]() |
Novus Biologicals
NBP15290120UL |
20 μL |
Each for $206.00
|
|
|||||
NBP152901
![]() |
Novus Biologicals
NBP152901 |
100 μL |
Each for $499.50
|
|
|||||
Description
BMP2K Polyclonal specifically detects BMP2K in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
BMP2K | |
Polyclonal | |
Purified | |
RUO | |
Q4W5H2 | |
55589 | |
Synthetic peptides corresponding to BMP2K(BMP2 inducible kinase) The peptide sequence was selected from the C terminal of BMP2K. Peptide sequence AQHQPSQQQASPEYLTSPQEFSPALVSYTSSLPAQVGTIMDSSYSANRSV. | |
Primary | |
74 kDa |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Protein Kinase, Stem Cell Signaling Pathway | |
BIKe, BMP2 inducible kinase, BMP-2-inducible protein kinase, DKFZp434K0614, DKFZp434P0116, EC 2.7.11.1 | |
BMP2K | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title