Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BMP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109450
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109450 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109450 Supplier Invitrogen™ Supplier No. PA5109450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YANDAAISEP ESVVSSLQGH RNFPTGTVPK WDSHIRAALS IERRKKRSTG VLLPLQNNEL PGAEYQYKKD EVWEERKPYK TLQAQAPEKS KNKKKQRKGP HRKSQTLQFD EQTLKKARRK QWIEPRNCAR RYLKVDFADI GWSEWIISPK SFDAYYCSGA CQFPMPKSLK PSNHATIQSI VRAVGVVPGI PEPCCVPEKM SSLSILFFDE NKNVVLKVYP NMTVESCACR.

The BMPs belong to the TGF-Beta superfamilyBMPs stimulate the production of bone matrix proteins and thereby alter stromal cell and osteoclast proliferation, a key step in bone marrow development. In addition to promoting ectopic bone and cartilage development, BMPs regulate the growth, differentiation, chemotaxis, proliferation, and apoptosis of various cell types, including mesenchymal cells, epithelial cells, hematopoietic cells, and neuronal cells. BMPs also play a role in dorsal/ventral patterning. BMP3, highly expressed in lung, ovary and small intestine, participates in cartilage and bone formation. BMP3 and BMP2 genes map to conserved regions between human and mouse.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMP3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BMP3
Gene Accession No. P12645, P49002, Q8BHE5
Gene Alias 9130206H07; 9530029I04Rik; BMP; Bmp3; Bmp-3; BMP3A; BMP-3A; Bone morphogenetic protein; Bone morphogenetic protein 3; bone morphogenetic protein 3 (osteogenic); bone morphogenetic protein 3A; bone morphogenetic protein-3; osteogenin; PBMP3
Gene Symbols BMP3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 243-472 of human BMP3 (NP_001192.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110075, 25667, 651
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.