Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ BMP5 ELISA Kit
Click to view available options
Quantity:
1 Kit
Description
BMP5 ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative measurement of bovine BMP5 in cell culture supernates, serum and bone tissue.
Specifications
Specifications
Description | bone morphogenetic protein 5 |
Cross Reactivity | Bovine |
Target Species | Bovine |
For Use With (Application) | Quantitative |
Gene ID (Entrez) | 507682 |
Quantity | 1 Kit |
Regulatory Status | RUO |
Storage Requirements | Store at 4°C for 6 months, at -20°C for 12 months. Avoid multiple freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction