Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BMP9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587966
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587966 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587966 Supplier Invitrogen™ Supplier No. PA587966
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SAGAGSHCQK TSLRVNFEDI GWDSWIIAPK EYEAYECKGG CFFPLADDVT PTKHAIVQTL VHLKFPTKVG KACCVPTKLS PISVLYKDDM GVPTLKYHYE GMSVAECGCR; Positive Samples: Mouse pancreas, Mouse skeletal muscle, Rat skeletal muscle.

The protein encoded by this gene is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in rodents suggest that this protein plays a role in the adult liver and in differentiation of cholinergic central nervous system neurons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMP9
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene GDF2
Gene Accession No. Q9WV56
Gene Alias BMP; BMP9; BMP-9; Bone morphogenetic protein; Bone morphogenetic protein 9; Gdf2; GDF-2; growth differentiation factor 2; growth/differentiation factor 2; HHT5
Gene Symbols GDF2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 320-429 of human GDF2 (NP_0572881).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12165, 290921
Target Species Mouse, Rat, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.