Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BMPER Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5118984
This item is not returnable.
View return policy
Description
BMPER Polyclonal Antibody for Western Blot, IHC (P)
This gene encodes a secreted protein that interacts with, and inhibits bone morphogenetic protein (BMP) function. It has been shown to inhibit BMP2- and BMP4-dependent osteoblast differentiation and BMP-dependent differentiation of the chondrogenic cells. Mutations in this gene are associated with a lethal skeletal disorder, diaphanospondylodysostosis.
Specifications
BMPER | |
Polyclonal | |
Unconjugated | |
Bmper | |
BMP-Binding Endothelial Regulator Protein; CRIM3 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
73230 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q8CJ69 | |
Bmper | |
Recombinant protein BMP Binding Endothelial Regulator. The antigen corresponds to amino acid range 40-298 of the target protein. | |
200 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction