Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BMPER Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5122195
This item is not returnable.
View return policy
Description
BMPER Polyclonal Antibody for Western Blot, IHC (P)
This gene encodes a secreted protein that interacts with, and inhibits bone morphogenetic protein (BMP) function. It has been shown to inhibit BMP2- and BMP4-dependent osteoblast differentiation and BMP-dependent differentiation of the chondrogenic cells. Mutations in this gene are associated with a lethal skeletal disorder, diaphanospondylodysostosis.
Specifications
BMPER | |
Polyclonal | |
Unconjugated | |
Bmper | |
BMP-Binding Endothelial Regulator Protein; CRIM3 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
73230 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q8CJ69 | |
Bmper | |
Recombinant protein BMP Binding Endothelial Regulator. The antigen corresponds to amino acid range 40-298 of the target protein. | |
20 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction